siRNA Details
| Gene Symbol: TNFRSF6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF104419.1 | - | 1 | 346 | ||
| AF134240.1 | - | 1 | 246 | |||
| AF217794.1 | - | 1 | 298 | |||
| AY124379.1 | - | 1 | 285 | |||
| AY358279.1 | - | 1 | 346 | |||
| BC017065.1 | - | 1 | 345 | 3886635 | ||
| BM759033.1 | - | 1 | 365 | |||

TNFRSF6B
Genomic Map