siRNA Details
| Gene Symbol: IER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003897.3 | NP_003888.2 | 1 | 130 | ||
| GenBank mRNA | AF039067.1 | - | 1 | 98 | ||
| AF083421.1 | - | 1 | 104 | |||
| AK222588.1 | - | 1 | 127 | |||
| AK314162.1 | - | 1 | 131 | |||
| BC000844.2 | - | 1 | 114 | 3457670 | ||
| BC005080.1 | - | 1 | 116 | 3617088 | ||
| BM915952.1 | - | 1 | 135 | |||
| BT006703.1 | - | 1 | 98 | |||
| DA893782.1 | - | 1 | 131 | |||
| S81914.1 | - | 1 | 116 | |||
| Y14551.1 | - | 1 | 109 | |||

IER3
Genomic Map