siRNA Details
| Gene Symbol: IER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003897.3 | NP_003888.2 | 1 | 218 | ||
| GenBank mRNA | AF083421.1 | - | 1 | 192 | ||
| AK222588.1 | - | 1 | 215 | |||
| AK314162.1 | - | 1 | 219 | |||
| BC000844.2 | - | 1 | 202 | 3457670 | ||
| BC005080.1 | - | 1 | 204 | 3617088 | ||
| BM915952.1 | - | 1 | 223 | |||
| BT006703.1 | - | 1 | 186 | |||
| DA893782.1 | - | 1 | 219 | |||
| S81914.1 | - | 1 | 204 | |||
| Y14551.1 | - | 1 | 197 | |||

IER3
Genomic Map