siRNA Details
| Gene Symbol: AP3D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001261826.1 | NP_001248755.1 | 32 | 3822 | ||
| NM_003938.6 | NP_003929.4 | 30 | 3636 | |||
| XM_006722932.2 | XP_006722995.1 | 32 | 3849 | |||
| XM_017027422.1 | XP_016882911.1 | 27 | 3119 | |||
| GenBank mRNA | AB208804.1 | - | 32 | 3806 | ||
| AF002163.1 | - | 3724 | ||||
| AK094460.1 | - | 6 | 2124 | |||
| AK128692.1 | - | 5 | 3206 | |||
| AK310099.1 | - | 7 | 836 | |||
| AK315899.1 | - | 8 | 811 | |||
| U91930.1 | - | 32 | 3499 | |||

AP3D1
Genomic Map