siRNA Details
| Gene Symbol: TRIM15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033229.2 | NP_150232.2 | 4, 5 | 1213 | ||
| XM_011514987.1 | XP_011513289.1 | 5, 6 | 452 | |||
| XM_011514988.2 | XP_011513290.1 | 4, 5 | 720 | |||
| GenBank mRNA | AF220132.1 | - | 4, 5 | 734 | ||
| AK290280.1 | - | 4, 5 | 919 | |||
| AK291853.1 | - | 4, 5 | 871 | |||
| BC038585.1 | - | 4, 5 | 814 | 5758860 | ||
| U34249.1 | - | 4, 5 | 1227 | |||

TRIM15
Genomic Map