siRNA Details
| Gene Symbol: ZNF766 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010851.2 | NP_001010851.1 | 3 | 294 | ||
| XM_011527460.1 | XP_011525762.1 | 3 | 621 | |||
| XM_017027440.1 | XP_016882929.1 | 3 | 489 | |||
| GenBank mRNA | AK024074.1 | - | 3 | 272 | ||
| AK097729.1 | - | 4 | 675 | |||
| BC109201.1 | - | 3 | 419 | |||
| BC136833.1 | - | 3 | 282 | 9020824 | ||
| BC136834.1 | - | 3 | 282 | 9020825 | ||
| BX538183.1 | - | 3 | 263 | |||
| BX648899.1 | - | 3 | 295 | |||

ZNF766
Genomic Map