siRNA Details
| Gene Symbol: TM4SF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003963.2 | NP_003954.2 | 1 | 113 | ||
| GenBank mRNA | AF027204.1 | - | 1 | 114 | ||
| BC069519.1 | - | 1 | 113 | 7262226 | ||
| BC093688.1 | - | 1 | 110 | 7939533 | ||
| BC117163.1 | - | 1 | 110 | 40125714 | ||
| CR456925.1 | - | 1 | 82 | |||
| CR541798.1 | - | 1 | 82 | |||

TM4SF5
Genomic Map