siRNA Details
| Gene Symbol: ATP6V1E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318063.1 | NP_001304992.1 | 5 | 1318 | ||
| XM_005264633.2 | XP_005264690.1 | 5 | 2049 | |||
| XM_005264634.2 | XP_005264691.1 | 4 | 1942 | |||
| XM_011533148.2 | XP_011531450.1 | 6 | 2748 | |||
| XM_011533149.2 | XP_011531451.1 | 5 | 2641 | |||
| XM_011533150.2 | XP_011531452.1 | 7 | 2949 | |||
| XM_011533151.2 | XP_011531453.1 | 6 | 2103 | |||
| XM_011533152.2 | XP_011531454.1 | 6 | 2820 | |||
| XM_011533153.2 | XP_011531455.1 | 5 | 1996 | |||
| XM_017005225.1 | XP_016860714.1 | 7 | 1869 | |||
| XM_017005226.1 | XP_016860715.1 | 5 | 2121 | |||
| GenBank mRNA | AB074759.1 | - | 2 | 979 | ||
| AK058055.1 | - | 2 | 752 | |||
| BC008981.1 | - | 2 | 979 | 3456620 | ||
| BC034808.1 | - | 2 | 1718 | 5167103 | ||
| HY189673.1 | - | 1 | 297 | |||

ATP6V1E2
Genomic Map