siRNA Details
| Gene Symbol: RPL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034996.2 | NP_001030168.1 | 2 | 215 | ||
| NM_003973.4 | NP_003964.3 | 2 | 151 | |||
| GenBank mRNA | AK222750.1 | - | 1 | 110 | ||
| AK223111.1 | - | 2 | 104 | |||
| AK290674.1 | - | 2 | 111 | |||
| AK292045.1 | - | 2 | 109 | |||
| AK300873.1 | - | 2 | 104 | |||
| BC000606.2 | - | 2 | 117 | 3346054 | ||
| BC005134.2 | - | 1 | 82 | 3928410 | ||
| BC009294.2 | - | 2 | 114 | 4123062 | ||
| BC016379.1 | - | 1 | 108 | |||
| BC019651.1 | - | 1 | 89 | 4454086 | ||
| BC022805.1 | - | 1 | 79 | |||
| BC029036.1 | - | 2 | 203 | 5183728 | ||
| BC071913.1 | - | 1 | 111 | 6671085 | ||
| BG564298.1 | - | 2 | 151 | |||
| BG776586.1 | - | 2 | 134 | |||
| CD298951.1 | - | 2 | 218 | |||
| D87735.1 | - | 2 | 99 | |||
| DQ118667.1 | - | 1 | 82 | |||
| L10376.1 | - | 1 | 87 | |||
| U16738.1 | - | 1 | 88 | |||
| U92982.1 | - | 1 | 15 | |||

RPL14
Genomic Map