siRNA Details
| Gene Symbol: ZNF835 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005850.2 | NP_001005850.2 | 2 | 594 | ||
| XM_005259382.2 | XP_005259439.1 | 2 | 597 | |||
| XM_005259383.3 | XP_005259440.1 | 2 | 629 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF835| Gene Symbol: ZNF835 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005850.2 | NP_001005850.2 | 2 | 594 | ||
| XM_005259382.2 | XP_005259439.1 | 2 | 597 | |||
| XM_005259383.3 | XP_005259440.1 | 2 | 629 | |||