siRNA Details
| Gene Symbol: GPRC5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003979.3 | NP_003970.1 | 2 | 1422 | ||
| GenBank mRNA | AF095448.1 | - | 2 | 871 | ||
| AF506289.1 | - | 2 | 879 | |||
| AK001761.1 | - | 2 | 1025 | |||
| AK122672.1 | - | 2 | 1662 | |||
| AK172760.1 | - | 1 | 392 | |||
| AK291171.1 | - | 2 | 857 | |||
| BC003665.2 | - | 2 | 866 | 2988011 | ||
More Information
Set Membership: |
|

GPRC5A
Genomic Map