siRNA Details
| Gene Symbol: SLC7A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001076785.2 | NP_001070253.1 | 4 | 703 | ||
| NM_003983.5 | NP_003974.3 | 3 | 616 | |||
| XM_011523433.1 | XP_011521735.1 | 3 | 718 | |||
| XM_011523434.1 | XP_011521736.1 | 3 | 783 | |||
| XM_011523438.1 | XP_011521740.1 | 2 | 1173 | |||
| XM_011523439.1 | XP_011521741.1 | 3 | 1302 | |||
| GenBank mRNA | AK310866.1 | - | 3 | 582 | ||
| AK311610.1 | - | 1 | 248 | |||
| BC028216.1 | - | 3 | 572 | 5214340 | ||
| BC113100.1 | - | 3 | 603 | |||
| CR749291.1 | - | 3 | 602 | |||
| CR749475.1 | - | 3 | 585 | |||
| D87432.1 | - | 3 | 651 | |||

SLC7A6
Genomic Map