siRNA Details
| Gene Symbol: COX19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031617.2 | NP_001026788.1 | 3 | 328 | ||
| GenBank mRNA | AK023185.1 | - | 3 | 263 | ||
| AY957566.1 | - | 3 | 278 | |||
| BC042434.1 | - | 3 | 238 | |||
| BC053372.1 | - | 3 | 245 | |||
| BC058158.1 | - | 3 | 260 | |||
| BC070383.1 | - | 3 | 274 | 6724444 | ||
| BC103632.1 | - | 3 | 304 | 6606556 | ||
| BC110420.1 | - | 3 | 266 | 5208612 | ||
| DB077585.1 | - | 3 | 328 | |||

COX19
Genomic Map