siRNA Details
| Gene Symbol: CARMIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138360.3 | NP_612369.3 | 9, 10 | 833 | ||
| XM_011537300.2 | XP_011535602.1 | 8, 9 | 492 | |||
| XM_017021751.1 | XP_016877240.1 | 5, 6 | 393 | |||
| GenBank mRNA | BU102125.1 | - | 5, 6 | 450 | ||

CARMIL3
Genomic Map