siRNA Details
| Gene Symbol: ZNF697 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080470.1 | NP_001073939.1 | 3 | 540 | ||
| XM_005271315.3 | XP_005271372.1 | 3 | 566 | |||
| XM_011542416.2 | XP_011540718.1 | 3 | 3545 | |||
| XM_017002783.1 | XP_016858272.1 | 3 | 801 | |||

ZNF697
Genomic Map