siRNA Details
| Gene Symbol: C12orf29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009894.2 | NP_001009894.2 | 6 | 867 | ||
| GenBank mRNA | AL137488.1 | - | 5 | 2775 | ||
| BC092422.1 | - | 6 | 809 | 30386540 | ||
| BX648026.1 | - | 5 | 703 | |||
| CR936776.1 | - | 6 | 856 | |||

C12orf29
Genomic Map