siRNA Details
| Gene Symbol: ZMYM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005095.2 | NP_005086.2 | 2, 3 | 169 | ||
| XM_005271328.4 | XP_005271385.1 | 2, 3 | 448 | |||
| XM_011542421.2 | XP_011540723.1 | 2, 3 | 95 | |||
| XM_011542423.2 | XP_011540725.1 | 3, 4 | 500 | |||
| XM_017002803.1 | XP_016858292.1 | 3, 4 | 222 | |||
| XM_017002804.1 | XP_016858293.1 | 2, 3 | 86 | |||
| GenBank mRNA | AK308320.1 | - | 2, 3 | 169 | ||
| BC127113.1 | - | 3, 4 | 175 | 40125110 | ||
| BG499626.1 | - | 2, 3 | 170 | |||
| CD653525.1 | - | 1, 2 | 63 | |||
| CR936741.1 | - | 1, 2 | 33 | |||

ZMYM4
Genomic Map