siRNA Details
| Gene Symbol: RIPPLY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171706.1 | NP_001165177.1 | 1 | 136 | ||
| NM_138382.2 | NP_612391.1 | 1 | 136 | |||
| GenBank mRNA | BC007652.1 | - | 1 | 122 | ||
| BC101998.1 | - | 1 | 136 | |||
| BC105691.1 | - | 1 | 136 | 40023299 | ||
| BC105692.1 | - | 1 | 136 | 40023298 | ||
| BC110436.1 | - | 1 | 136 | 40023301 | ||
| BC127250.1 | - | 1 | 136 | 40023300 | ||
| BX106639.1 | - | 1 | 172 | |||

RIPPLY1
Genomic Map