siRNA Details
| Gene Symbol: VAPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004738.4 | NP_004729.1 | 3 | 574 | ||
| GenBank mRNA | AF086628.1 | - | 3 | 380 | ||
| AF160212.1 | - | 3 | 272 | |||
| AK023241.1 | - | 3 | 571 | |||
| AK123538.1 | - | 3 | 577 | |||
| AY358464.1 | - | 3 | 466 | |||
| BC001712.1 | - | 4 | 411 | 3543354 | ||
| BT009811.1 | - | 3 | 233 | |||

VAPB
Genomic Map