siRNA Details
| Gene Symbol: RCSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322923.1 | NP_001309852.1 | 5 | 673 | ||
| NM_001322924.1 | NP_001309853.1 | 4 | 601 | |||
| NM_052862.3 | NP_443094.3 | 6 | 719 | |||
| GenBank mRNA | AF545852.1 | - | 6 | 863 | ||
| AK025425.1 | - | 1 | 58 | |||
| AL137762.1 | - | 6 | 640 | |||
| AL833211.1 | - | 4 | 409 | |||
| AY530954.1 | - | 5 | 532 | |||
| BC072399.1 | - | 11 | ||||
| BC101536.1 | - | 6 | 556 | 8069042 | ||
| BC101562.1 | - | 6 | 556 | 8069068 | ||
| BC143428.1 | - | 5 | 466 | 9051936 | ||

RCSD1
Genomic Map