siRNA Details
| Gene Symbol: TMEM132C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136103.2 | NP_001129575.2 | 9 | 3156 | ||
| XM_011538998.2 | XP_011537300.1 | 9 | 3482 | |||
| GenBank mRNA | AK126715.1 | - | 6 | 2025 | ||
| AL122107.1 | - | 1 | 239 | |||
| AL831921.1 | - | 6 | 2003 | |||
| AL832160.1 | - | 4 | 1630 | |||

TMEM132C
Genomic Map