siRNA Details
| Gene Symbol: PTTG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282382.1 | NP_001269311.1 | 2 | 574 | ||
| NM_001282383.1 | NP_001269312.1 | 3 | 428 | |||
| NM_004219.3 | NP_004210.1 | 3 | 267 | |||
| GenBank mRNA | AF075242.1 | - | 2 | 191 | ||
| AF095287.1 | - | 3 | 207 | |||
| AJ223953.1 | - | 2 | 276 | |||
| AK312209.1 | - | 3 | 245 | |||
| BC101834.1 | - | 3 | 200 | 8069340 | ||
| BC108894.1 | - | 2 | 171 | |||
| BC108895.1 | - | 2 | 171 | |||
| BC112071.1 | - | 3 | 200 | 8327539 | ||
| BC113842.1 | - | 2 | 171 | |||
| BC113896.1 | - | 2 | 171 | |||
| BC128193.1 | - | 2 | 171 | 40113885 | ||
| BC128194.1 | - | 2 | 171 | 40113886 | ||
| BI827526.1 | - | 2 | 574 | |||
| BM990391.1 | - | 4 | 516 | |||
| BU182149.1 | - | 3 | 423 | |||
| CR457135.1 | - | 2 | 171 | |||
| CR541685.1 | - | 2 | 171 | |||
| HY006026.1 | - | 3 | 268 | |||

PTTG1
Genomic Map