siRNA Details
| Gene Symbol: GORAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320252.1 | NP_001307181.1 | 7 | 1348 | ||
| NM_152281.2 | NP_689494.2 | 5 | 1048 | |||
| NR_027397.1 | 5 | 1052 | ||||
| XM_006711628.3 | XP_006711691.1 | 7 | 1323 | |||
| XM_011510149.2 | XP_011508451.1 | 5 | 985 | |||
| XM_011510150.2 | XP_011508452.1 | 6 | 1226 | |||
| XM_011510151.1 | XP_011508453.1 | 5 | 975 | |||
| XM_017002807.1 | XP_016858296.1 | 7 | 1289 | |||
| GenBank mRNA | AF143889.1 | - | 2 | 444 | ||
| AK021814.1 | - | 5 | 1041 | |||
| AK057661.1 | - | 5 | 858 | |||
| BC047476.1 | - | 7 | 1319 | 5296051 | ||

GORAB
Genomic Map