siRNA Details
| Gene Symbol: ZFP62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172638.1 | NP_001166109.1 | 2 | 2536 | ||
| NM_152283.4 | NP_689496.4 | 3 | 2609 | |||
| XM_017009710.1 | XP_016865199.1 | 3 | 2639 | |||
| XM_017009711.1 | XP_016865200.1 | 3 | 2687 | |||
| XM_017009712.1 | XP_016865201.1 | 4 | 2712 | |||
| XM_017009713.1 | XP_016865202.1 | 4 | 2760 | |||
| XM_017009714.1 | XP_016865203.1 | 3 | 2693 | |||
| XM_017009715.1 | XP_016865204.1 | 2 | 2646 | |||
| XM_017009717.1 | XP_016865206.1 | 2 | 2609 | |||
| GenBank mRNA | AK002206.1 | - | 1 | 1092 | ||
| AK091550.1 | - | 2 | 1366 | |||
| AK295714.1 | - | 3 | 2599 | |||
| AK303450.1 | - | 2 | 2527 | |||
| AL832408.1 | - | 1 | 2696 | |||
| BC140761.1 | - | 1 | 1915 | 9021629 | ||
| BC171780.1 | - | 1 | 1915 | |||

ZFP62
Genomic Map