siRNA Details
| Gene Symbol: ASB16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080863.4 | NP_543139.4 | 1 | 117 | ||
| GenBank mRNA | AK054727.1 | - | 1 | 117 | ||
| AK314595.1 | - | 1 | 109 | |||
| AY557346.1 | - | 1 | 33 | |||
| BC075088.2 | - | 1 | 66 | 30915388 | ||
| BC118584.1 | - | 1 | 99 | 40084995 | ||
| BC118585.1 | - | 1 | 99 | 40084992 | ||

ASB16
Genomic Map