siRNA Details
| Gene Symbol: DNER | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139072.3 | NP_620711.3 | 9 | 1707 | ||
| GenBank mRNA | AF442487.1 | - | 9 | 1620 | ||
| AK289553.1 | - | 9 | 1665 | |||
| AK304596.1 | - | 7 | 1057 | |||
| AK304631.1 | - | 8 | 1059 | |||
| AK308576.1 | - | 5 | 410 | |||
| AL137311.1 | - | 2 | 105 | |||
| AY358891.1 | - | 9 | 1670 | |||
| BC024766.2 | - | 5 | 713 | |||
| BC035009.1 | - | 9 | 1697 | 4820343 | ||

DNER
Genomic Map