siRNA Details
| Gene Symbol: REEP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138393.1 | NP_612402.1 | 1, 2 | 227 | ||
| GenBank mRNA | AK293919.1 | - | 1, 2 | 200 | ||
| AK315744.1 | - | 1, 2 | 202 | |||
| AX775999.1 | - | 1, 2 | 227 | |||
| AY562244.1 | - | 1, 2 | 123 | |||
| BC008201.1 | - | 1, 2 | 227 | 3865853 | ||

REEP6
Genomic Map