siRNA Details
| Gene Symbol: COQ10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099337.1 | NP_001092807.1 | 3 | 355 | ||
| NM_144576.3 | NP_653177.3 | 3 | 599 | |||
| GenBank mRNA | AK057003.1 | - | 3 | 515 | ||
| AK057014.1 | - | 2 | 316 | |||
| AY358861.1 | - | 3 | 287 | |||
| BC002435.2 | - | 2 | 176 | |||
| BC026922.1 | - | 3 | 355 | |||
| BC047444.1 | - | 3 | 327 | |||
| BC073923.1 | - | 3 | 483 | 5188138 | ||

COQ10A
Genomic Map