siRNA Details
| Gene Symbol: ZNF235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004234.4 | NP_004225.3 | 4, 5 | 342 | ||
| GenBank mRNA | AK300317.1 | - | 4, 5 | 342 | ||
| AK315242.1 | - | 4, 5 | 317 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF235| Gene Symbol: ZNF235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004234.4 | NP_004225.3 | 4, 5 | 342 | ||
| GenBank mRNA | AK300317.1 | - | 4, 5 | 342 | ||
| AK315242.1 | - | 4, 5 | 317 | |||