siRNA Details
| Gene Symbol: GTF3C3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012086.4 | NP_036218.1 | 14 | 2029 | ||
| XM_005246965.4 | XP_005247022.1 | 14 | 2033 | |||
| GenBank mRNA | AF133123.1 | - | 14 | 1965 | ||
| AK025560.1 | - | 14 | 1943 | |||
| AK027050.1 | - | 8 | 980 | |||
| AK314718.1 | - | 14 | 1970 | |||
| AL137453.1 | - | 11 | 1371 | |||
| BC015995.1 | - | 6 | 728 | |||
| BC043347.1 | - | 14 | 1884 | 5498609 | ||

GTF3C3
Genomic Map