siRNA Details
| Gene Symbol: MVB12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304547.1 | NP_001291476.1 | 8 | 882 | ||
| NM_138401.3 | NP_612410.1 | 7 | 955 | |||
| GenBank mRNA | AK057057.1 | - | 8 | 1440 | ||
| AK289487.1 | - | 7 | 755 | |||
| BC007883.2 | - | 7 | 691 | 3688605 | ||
| BC011840.2 | - | 7 | 699 | 4053084 | ||
| BY996463.1 | - | 5 | 516 | |||

MVB12A
Genomic Map