siRNA Details
| Gene Symbol: SYCE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143763.1 | NP_001137235.1 | 10 | 780 | ||
| NM_001143764.1 | NP_001137236.1 | 10 | 780 | |||
| NM_130784.2 | NP_570140.1 | 10 | 703 | |||
| GenBank mRNA | AK314978.1 | - | 10 | 730 | ||
| AY027807.1 | - | 10 | 703 | |||
| AY027808.1 | - | 7 | 1639 | |||
| BC034821.1 | - | 10 | 730 | 5164386 | ||
| DB303602.1 | - | 4 | 526 | |||

SYCE1
Genomic Map