siRNA Details
| Gene Symbol: UBE4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204077.1 | NP_001191006.1 | 14 | 2349 | ||
| NM_004788.3 | NP_004779.2 | 14 | 2370 | |||
| GenBank mRNA | AK296369.1 | - | 14 | 2296 | ||
| AK302328.1 | - | 5 | 1095 | |||
| AK307737.1 | - | 9 | 1546 | |||
| BC111417.1 | - | 14 | 2323 | 40038776 | ||
| BC112367.1 | - | 14 | 2326 | 40038767 | ||
| D50916.1 | - | 14 | 2307 | |||

UBE4A
Genomic Map