siRNA Details
| Gene Symbol: ZNF264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003417.4 | NP_003408.1 | 4 | 801 | ||
| XM_011527522.2 | XP_011525824.1 | 3 | 705 | |||
| GenBank mRNA | AB007872.1 | - | 4 | 750 | ||
| AK292504.1 | - | 4 | 801 | |||
| BC115410.2 | - | 4 | 443 | |||
| BC115411.1 | - | 5 | 483 | 40078437 | ||

ZNF264
Genomic Map