siRNA Details
| Gene Symbol: SCAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290229.1 | NP_001277158.1 | 5 | 687 | ||
| NM_004866.5 | NP_004857.4 | 6 | 765 | |||
| XM_011543727.2 | XP_011542029.1 | 6 | 765 | |||
| GenBank mRNA | AF005037.1 | - | 6 | 639 | ||
| AF038966.1 | - | 6 | 686 | |||
| AK001541.1 | - | 1 | 95 | |||
| AK225172.1 | - | 6 | 654 | |||
| AK290225.1 | - | 6 | 615 | |||
| AK298146.1 | - | 4 | 398 | |||
| AK304452.1 | - | 5 | 579 | |||
| BC015065.1 | - | 6 | 658 | 4105068 | ||
| BC034048.1 | - | 6 | 619 | 4814951 | ||
| CR542286.1 | - | 6 | 596 | |||

SCAMP1
Genomic Map