siRNA Details
| Gene Symbol: TP53I3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206802.2 | NP_001193731.1 | 2, 3 | 420 | ||
| NM_004881.4 | NP_004872.2 | 2, 3 | 1304 | |||
| NM_147184.3 | NP_671713.1 | 3, 4 | 905 | |||
| XM_006712150.2 | XP_006712213.1 | 3, 4 | 905 | |||
| GenBank mRNA | AF010309.1 | - | 2, 3 | 942 | ||
| AK223382.1 | - | 2, 3 | 1141 | |||
| AY371700.1 | - | 2, 3 | 415 | |||
| BC000474.2 | - | 3, 4 | 475 | 2961571 | ||
| BC018819.2 | - | 3, 4 | 475 | |||
| BT007149.1 | - | 2, 3 | 415 | |||

TP53I3
Genomic Map