siRNA Details
| Gene Symbol: SPAG6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253854.1 | NP_001240783.1 | 10 | 1464 | ||
| NM_001253855.1 | NP_001240784.1 | 6 | 1195 | |||
| NM_012443.3 | NP_036575.1 | 7 | 1109 | |||
| NM_172242.2 | NP_758442.1 | 7 | 1109 | |||
| XM_005252645.1 | XP_005252702.1 | 7 | 1138 | |||
| XM_005252646.1 | XP_005252703.1 | 6 | 964 | |||
| GenBank mRNA | AF079363.1 | - | 7 | 1066 | ||
| AK289903.1 | - | 7 | 1058 | |||
| AK302194.1 | - | 10 | 1439 | |||
| AK310247.1 | - | 6 | 1148 | |||
| AL080136.1 | - | 7 | 1057 | |||
| BC030585.2 | - | 7 | 1057 | 4822996 | ||
| CR533552.1 | - | 7 | 942 | |||
| HM370395.1 | - | 2 | 120 | |||

SPAG6
Genomic Map