siRNA Details
| Gene Symbol: ACY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000666.2 | NP_000657.1 | 9, 10 | 981 | ||
| NM_001198895.1 | NP_001185824.1 | 9, 10 | 976 | |||
| NM_001198896.1 | NP_001185825.1 | 7, 8 | 765 | |||
| NM_001198898.1 | NP_001185827.1 | 8, 9 | 876 | |||
| GenBank mRNA | AK298274.1 | - | 9, 10 | 730 | ||
| BC000545.2 | - | 9, 10 | 725 | 3162222 | ||
| BC003023.1 | - | 9, 10 | 718 | 2821665 | ||
| BC014112.2 | - | 9, 10 | 701 | 4543442 | ||
| CD014023.1 | - | 7, 8 | 606 | |||
| CD014027.1 | - | 8, 9 | 717 | |||
| D14524.1 | - | 9, 10 | 715 | |||
| D16307.1 | - | 9, 10 | 713 | |||
| EU794586.1 | - | 9, 10 | 701 | |||
| KU177890.1 | - | 8, 9 | 657 | |||
| L07548.1 | - | 9, 10 | 718 | |||
| Gene Symbol: ABHD14A-ACY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316331.1 | NP_001303260.1 | 11, 12 | 1182 | ||
| GenBank mRNA | AK298086.1 | - | 11, 12 | 1158 | ||

ACY1
Genomic Map