siRNA Details
| Gene Symbol: SLC25A44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286184.1 | NP_001273113.1 | 2 | 927 | ||
| NM_014655.3 | NP_055470.1 | 2 | 927 | |||
| XM_006711657.3 | XP_006711720.1 | 3 | 1309 | |||
| XM_011510180.1 | XP_011508482.1 | 4 | 1660 | |||
| XM_011510181.1 | XP_011508483.1 | 3 | 1485 | |||
| XM_017002904.1 | XP_016858393.1 | 4 | 1660 | |||
| XM_017002905.1 | XP_016858394.1 | 3 | 1485 | |||
| XM_017002906.1 | XP_016858395.1 | 3 | 1306 | |||
| XM_017002907.1 | XP_016858396.1 | 2 | 775 | |||
| XM_017002908.1 | XP_016858397.1 | 3 | 1485 | |||
| GenBank mRNA | AB007915.3 | - | 2 | 730 | ||
| AK074912.1 | - | 2 | 772 | |||
| AK075002.1 | - | 2 | 747 | |||
| AK294524.1 | - | 3 | 674 | |||
| AK299747.1 | - | 3 | 429 | |||
| AK307968.1 | - | 4 | 539 | |||
| BC008843.2 | - | 2 | 751 | 4136523 | ||
| BC039854.1 | - | 2 | 730 | 6061399 | ||

SLC25A44
Genomic Map