siRNA Details
Gene Symbol: KIAA0391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256678.1 | NP_001243607.1 | 6 | 2134 | ||
NM_001256679.1 | NP_001243608.1 | 7 | 1629 | |||
NM_001256680.1 | NP_001243609.1 | 6 | 946 | |||
NM_001256681.1 | NP_001243610.1 | 5 | 898 | |||
NM_014672.3 | NP_055487.2 | 7 | 2182 | |||
XM_005268237.3 | XP_005268294.1 | 7 | 2081 | |||
XM_011537409.2 | XP_011535711.1 | 7 | 2008 | |||
XM_011537410.2 | XP_011535712.1 | 7 | 2066 | |||
XM_017021835.1 | XP_016877324.1 | 7 | 1954 | |||
XM_017021836.1 | XP_016877325.1 | 6 | 2018 | |||
XM_017021837.1 | XP_016877326.1 | 6 | 2017 | |||
GenBank mRNA | AB002389.1 | - | 6 | 1884 | ||
AK301931.1 | - | 7 | 1403 | |||
AK303508.1 | - | 6 | 745 | |||
AK304066.1 | - | 6 | 1891 | |||
BC032221.1 | - | 7 | 1933 | 5206545 | ||
BC044580.1 | - | 7 | 1951 | 4822711 | ||
BQ063741.1 | - | 5 | 763 | |||
BX161394.1 | - | 7 | 1914 | |||
BX161487.1 | - | 6 | 776 | |||
DT217089.1 | - | 3 | 402 |
