siRNA Details
| Gene Symbol: KIAA0100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014680.3 | NP_055495.2 | 26, 27 | 5058 | ||
| XM_005258073.2 | XP_005258130.1 | 26, 27 | 5050 | |||
| XM_017025423.1 | XP_016880912.1 | 25, 26 | 4907 | |||
| GenBank mRNA | BC120873.1 | - | 26, 27 | 4917 | 40075102 | |
| CR933634.1 | - | 27, 28 | 4920 | |||
| D43947.3 | - | 26, 27 | 5030 | |||
| DB152098.1 | - | 3, 4 | 516 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001002004, X81632, BC075633, BC040464, AK122206, and others.

KIAA0100
Genomic Map