siRNA Details
| Gene Symbol: ULK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142610.1 | NP_001136082.1 | 6 | 839 | ||
| NM_014683.3 | NP_055498.3 | 6 | 839 | |||
| XM_011524087.2 | XP_011522389.1 | 2 | 136 | |||
| XM_017025424.1 | XP_016880913.1 | 6 | 727 | |||
| XM_017025425.1 | XP_016880914.1 | 6 | 727 | |||
| XM_017025426.1 | XP_016880915.1 | 6 | 727 | |||
| XM_017025427.1 | XP_016880916.1 | 5 | 240 | |||
| XM_017025428.1 | XP_016880917.1 | 5 | 240 | |||
| GenBank mRNA | AB014523.1 | - | 6 | 830 | ||
| AK098316.1 | - | 5 | 464 | |||
| AK314345.1 | - | 6 | 364 | |||
| BC034988.1 | - | 7 | 404 | 5268025 | ||

ULK2
Genomic Map