siRNA Details
| Gene Symbol: TMEM63A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014698.2 | NP_055513.2 | 19 | 2352 | ||
| XM_006711841.3 | XP_006711904.1 | 17 | 1845 | |||
| XM_011544328.2 | XP_011542630.1 | 19 | 2204 | |||
| XM_011544329.2 | XP_011542631.1 | 19 | 3471 | |||
| XM_011544330.2 | XP_011542632.1 | 18 | 2161 | |||
| XM_011544331.2 | XP_011542633.1 | 18 | 1948 | |||
| XM_011544332.2 | XP_011542634.1 | 18 | 1983 | |||
| XM_017002916.1 | XP_016858405.1 | 18 | 2435 | |||
| GenBank mRNA | AB018335.1 | - | 19 | 1980 | ||
| AK022502.1 | - | 19 | 2001 | |||
| AK222934.1 | - | 19 | 1982 | |||
| AK222944.1 | - | 19 | 1982 | |||
| BC030245.1 | - | 19 | 1985 | 5216414 | ||

TMEM63A
Genomic Map