siRNA Details
| Gene Symbol: SECISBP2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193489.1 | NP_001180418.1 | 12 | 1898 | ||
| NM_014701.3 | NP_055516.2 | 11 | 1763 | |||
| GenBank mRNA | AK307480.1 | - | 12 | 1581 | ||
| BC033001.2 | - | 12 | 1833 | 4825784 | ||
| D87445.3 | - | 11 | 1633 | |||

SECISBP2L
Genomic Map