siRNA Details
| Gene Symbol: USP34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014709.3 | NP_055524.3 | 80 | 10116 | ||
| GenBank mRNA | AB011142.2 | - | 81 | 10171 | ||
| AB018272.1 | - | 23 | 3045 | |||
| AJ586138.1 | - | 78 | 9641 | |||
| AK001193.1 | - | 20 | 2707 | |||
| AK022637.1 | - | 10 | 1156 | |||
| AK023171.1 | - | 12 | 1393 | |||
| AL050092.1 | - | 15 | 2212 | |||
| AL050376.1 | - | 1 | 632 | |||
| BC022783.1 | - | 17 | 2415 | |||
| BC062325.1 | - | 8 | 870 | |||
| BC107761.1 | - | 7 | 781 | |||
| Gene Symbol: AHSA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321305.1 | NP_001308234.1 | 6 | 2835 | ||
| XM_011532541.2 | XP_011530843.1 | 8 | 3187 | |||
| XM_011532543.2 | XP_011530845.1 | 9 | 3471 | |||
| XM_011532545.2 | XP_011530847.1 | 8 | 1895 | |||
| XM_017003377.1 | XP_016858866.1 | 7 | 1793 | |||
| XM_017003378.1 | XP_016858867.1 | 9 | 3146 | |||
| GenBank mRNA | AK091998.1 | - | 1 | 2363 | ||

USP34
Genomic Map