siRNA Details
| Gene Symbol: PTDSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014754.2 | NP_055569.1 | 4 | 528 | ||
| GenBank mRNA | AK293036.1 | - | 4 | 512 | ||
| BC004192.2 | - | 4 | 478 | 3509418 | ||
| BC004390.1 | - | 4 | 478 | 3634879 | ||
| D14694.1 | - | 4 | 475 | |||

PTDSS1
Genomic Map