siRNA Details
| Gene Symbol: URB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001314021.1 | NP_001300950.1 | 10 | 4688 | ||
| NM_014777.3 | NP_055592.2 | 10 | 4657 | |||
| XM_005273360.3 | XP_005273417.1 | 9 | 4515 | |||
| GenBank mRNA | AK291013.1 | - | 10 | 4657 | ||
| BC114559.1 | - | 10 | 4610 | 40082897 | ||
| BC114963.1 | - | 10 | 4579 | 40082900 | ||
| D50923.2 | - | 10 | 4639 | |||

URB2
Genomic Map