siRNA Details
| Gene Symbol: NUP58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008564.1 | NP_001008564.1 | 8 | 1096 | ||
| NM_014089.3 | NP_054808.1 | 9 | 1132 | |||
| XM_005266600.3 | XP_005266657.1 | 9 | 1476 | |||
| XM_011535328.1 | XP_011533630.1 | 9 | 1476 | |||
| XM_011535329.2 | XP_011533631.1 | 10 | 1562 | |||
| XM_017020880.1 | XP_016876369.1 | 11 | 1644 | |||
| XM_017020881.1 | XP_016876370.1 | 9 | 1537 | |||
| GenBank mRNA | AB007870.1 | - | 9 | 1029 | ||
| AK123955.1 | - | 10 | 966 | |||
| AK302946.1 | - | 8 | 991 | |||
| AK303409.1 | - | 8 | 900 | |||
| BC001104.2 | - | 9 | 1004 | 3504343 | ||

NUP58
Genomic Map