siRNA Details
| Gene Symbol: PLPPR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166252.1 | NP_001159724.1 | 4 | 765 | ||
| NM_014839.4 | NP_055654.2 | 4 | 765 | |||
| XM_011542498.2 | XP_011540800.1 | 4 | 713 | |||
| GenBank mRNA | AB007924.3 | - | 4 | 761 | ||
| AF357013.1 | - | 4 | 538 | |||
| AF541281.1 | - | 4 | 772 | |||
| AK291369.1 | - | 4 | 565 | |||
| AK300361.1 | - | 4 | 509 | |||
| AL834390.1 | - | 3 | 278 | |||
| AY304518.1 | - | 4 | 713 | |||
| BC042963.1 | - | 4 | 583 | |||

PLPPR4
Genomic Map