siRNA Details
| Gene Symbol: REC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048205.1 | NP_001041670.1 | 3 | 758 | ||
| NM_005132.2 | NP_005123.2 | 4 | 657 | |||
| XM_011537421.1 | XP_011535723.1 | 3 | 930 | |||
| XM_017021841.1 | XP_016877330.1 | 3 | 727 | |||
| GenBank mRNA | AF006264.1 | - | 3 | 230 | ||
| AK001435.1 | - | 4 | 657 | |||
| AK291191.1 | - | 4 | 789 | |||
| AL832973.1 | - | 3 | 648 | |||
| BC004159.2 | - | 3 | 314 | 2822680 | ||
| BC010887.1 | - | 3 | 691 | 3535425 | ||
| CR457263.1 | - | 3 | 230 | |||

REC8
Genomic Map